Learn More
Medchemexpress LLC Natural methyl nonyl ketone (2-undecanone) | 112-12-9 | MFCD00009583 | 99.5% | 170.30 g/mol | C11H22O | 100 G
SDP
Catalog No. 5000301261
Encompass_Preferred
2-Undecanone (C11H22O) is a volatile organic ketone used as a research-grade biochemical reagent. It inhibits the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has been reported to inhibit lung tumorigenesis. It is supplied at high purity for analytical and mechanistic laboratory studies.
  • Inhibits DnaKJE-ClpB chaperone-dependent refolding, enabling chaperone mechanism studies.
  • Reported to inhibit lung tumorigenesis, useful for cancer pathway research.
  • High purity suitable for analytical and mechanistic experiments.
  • Volatile liquid compatible with common organic solvents for formulation and assays.
  • Stable under recommended storage; solutions storable at -20°C to -80°C for extended periods.
Catalog No. 50-003-01261 Supplier Medchemexpress LLC Supplier No. HYW016969100G
2-Undecanone (C11H22O) is a volatile organic ketone used as a research-grade biochemical reagent. It inhibits the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has been reported to inhibit lung tumorigenesis. It is supplied at high purity for analytical and mechanistic laboratory studies.
  • Inhibits DnaKJE-ClpB chaperone-dependent refolding, enabling chaperone mechanism studies.
  • Reported to inhibit lung tumorigenesis, useful for cancer pathway research.
  • High purity suitable for analytical and mechanistic experiments.
  • Volatile liquid compatible with common organic solvents for formulation and assays.
  • Stable under recommended storage; solutions storable at -20°C to -80°C for extended periods.