Learn More
Trajan Scientific PEEK Tubing, Natural PEEK Tubing - 1/8" OD x 060" ID x 100FT, 3600 PSI
Catalog No. 501385126
Encompass_Preferred
PEEK Tubing, Natural PEEK Tubing - 1/8" OD x .060" ID x 100FT, 3600 PSI. PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting, and is not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel. The polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample. Note however that dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.
Catalog No. 50-138-5126 Supplier Trajan Scientific Supplier No. VAL.TPK260100
PEEK Tubing, Natural PEEK Tubing - 1/8" OD x .060" ID x 100FT, 3600 PSI. PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting, and is not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel. The polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample. Note however that dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.