Learn More
SKS SCIENCE Natural Polypro Hinge Top Vials, 1 1/2 oz., 48 per Bag
Catalog No. NC3726057
Encompass
Natural plastic lab vials with hinged lids are available here in cylinder styles and large base styles to meet your laboratory needs. The polypropylene construction makes these plastic lab vials durable and provides excellent moisture barriers and good acid, heat, aldehyde, base, ester, and ketone resistances. The secure hinged lids are permanently attached, ensuring no loss of the lids in fast pace laboratories or field collection.
Catalog No. NC3726057 Supplier SKS SCIENCE Supplier No. 074121
Natural plastic lab vials with hinged lids are available here in cylinder styles and large base styles to meet your laboratory needs. The polypropylene construction makes these plastic lab vials durable and provides excellent moisture barriers and good acid, heat, aldehyde, base, ester, and ketone resistances. The secure hinged lids are permanently attached, ensuring no loss of the lids in fast pace laboratories or field collection.