Learn More
Apexbio Technology LLC DiscoveryProbe™ Natural Product Library Plus, 250uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Catalog No. 501998480
Encompass_Preferred
DiscoveryProbe™ Natural Product Library Plus is a unique collection of 1170 natural products for high throughput screening (HTS) and high content screening (HCS). Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8480 Supplier Apexbio Technology LLC Supplier No. L1039P.25
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe™ Natural Product Library Plus is a unique collection of 1170 natural products for high throughput screening (HTS) and high content screening (HCS). Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.