Learn More
Enzo Life Sciences SCREEN-WELL Natural Product library (500 µl/well)
SDP
Supplier:  Enzo Life Sciences BML28650500
Encompass_Preferred
Natural products are an unsurpassed source of chemical diversity and an ideal starting point for any screening program for pharmacologically active small molecules. Historically, natural products have been the most successful source of new drugs. The SCREEN-WELL Natural Product library offers over 500 isolated natural compounds of known structure and includes many compounds from ethnopharmacology. Compounds are provided in solution in DMSO. Individual compounds or subsets can be resupplied in gram quantities. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0225
Only null left
Add to Cart
Request a Quote