Learn More
US Plastic Corporation Natural Standard Cover for 100, 150, 165 & 170 Gallon (30" Dia.) Tamco® Tanks
SDP
Catalog No. NC3918736
Encompass
This standard cover for cylindrical tanks fits most 30" and 31" diameter Tamco® tanks. The approximate inside diameter is 32-11/16" and has a 5/32" wall thickness. These covers are loose fitting and fit Tamco® tanks 4158, 4160, 4162, 4163 5208, and 5210. A drawing is required for custom sizes and special fitting placement. Please contact our quotes department at 1-800-335-6809 or email your drawing to quotes@usplastic.com.Made of a UV stabilized LLDPENot for fuels, oils, or wetting agentsMaximum temperature: 140°FMeets FDA standardsCovers sold separatelyDimensions are approximate outside dimensions
Catalog No. NC3918736 Supplier US Plastic Corporation Supplier No. 4210
This standard cover for cylindrical tanks fits most 30" and 31" diameter Tamco® tanks. The approximate inside diameter is 32-11/16" and has a 5/32" wall thickness. These covers are loose fitting and fit Tamco® tanks 4158, 4160, 4162, 4163 5208, and 5210. A drawing is required for custom sizes and special fitting placement. Please contact our quotes department at 1-800-335-6809 or email your drawing to quotes@usplastic.com.Made of a UV stabilized LLDPENot for fuels, oils, or wetting agentsMaximum temperature: 140°FMeets FDA standardsCovers sold separatelyDimensions are approximate outside dimensions