Learn More
Medchemexpress LLC HY-112473 100mg Medchemexpress, NAV-2729 CAS:419547-11-8 Purity:98%
SDP
Catalog No. 501637594
Encompass_Preferred
Medchemexpress,HY-112473 100mg NAV-2729 CAS:419547-11-8 Formula:C25H17ClN4O3 NAV-2729 is a dual Arf1/Arf6 activation inhibitor. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7594 Supplier Medchemexpress LLC Supplier No. HY112473100MG
Medchemexpress,HY-112473 100mg NAV-2729 CAS:419547-11-8 Formula:C25H17ClN4O3 NAV-2729 is a dual Arf1/Arf6 activation inhibitor. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.