Learn More
Medchemexpress LLC HY-112473 5mg Medchemexpress, NAV-2729 CAS:419547-11-8 Purity:>98%
SDP
Catalog No. 501969096
Encompass_Preferred
Medchemexpress, HY-112473 5mg NAV-2729 CAS:419547-11-8 NAV-2729 is a dual Arf1/Arf6 activation inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9096 Supplier Medchemexpress LLC Supplier No. HY1124735MG
Medchemexpress, HY-112473 5mg NAV-2729 CAS:419547-11-8 NAV-2729 is a dual Arf1/Arf6 activation inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.