Learn More
Medchemexpress LLC NaV1.7 inhibitor-1 | 1494585-79-3 | 99.2% | 100 MG
SDP
Catalog No. 5000234118
Encompass_Preferred
NaV1.7 inhibitor-1 is a potent research-grade small molecule inhibitor of the voltage-gated sodium channel NaV1.7, reported with sub-nanomolar potency in cellular assays. Supplied as a high-purity solid, it is intended for in vitro pharmacology and electrophysiology studies and is for research use only.
  • High potency against human NaV1.7 (IC50 = 0.6 nM).
  • Selective over NaV1.5 (~80-fold selectivity).
  • High purity (99.17% by LCMS).
  • Available in multiple pack sizes, including 100 MG solid and 10 mM DMSO solutions.
  • Molecular formula C23H30FNO4S, molecular weight ~435.6 g/mol.
  • Intended for laboratory research use only.
Catalog No. 50-002-34118 Supplier Medchemexpress LLC Supplier No. HY119934100MG
NaV1.7 inhibitor-1 is a potent research-grade small molecule inhibitor of the voltage-gated sodium channel NaV1.7, reported with sub-nanomolar potency in cellular assays. Supplied as a high-purity solid, it is intended for in vitro pharmacology and electrophysiology studies and is for research use only.
  • High potency against human NaV1.7 (IC50 = 0.6 nM).
  • Selective over NaV1.5 (~80-fold selectivity).
  • High purity (99.17% by LCMS).
  • Available in multiple pack sizes, including 100 MG solid and 10 mM DMSO solutions.
  • Molecular formula C23H30FNO4S, molecular weight ~435.6 g/mol.
  • Intended for laboratory research use only.