Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Navarixin | 473727-83-2 | 99.3% | 397.42 g/mol | C21H23N3O5 | 10 MG
SDP

Catalog No. 5000246742
Encompass_Preferred

Navarixin is an orally active, allosteric antagonist of CXCR1 and CXCR2 used as a research compound to study chemokine receptor signaling, neutrophil recruitment, and inflammatory responses in vitro and in vivo.

  • Potent, allosteric antagonist of CXCR1 and CXCR2.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High purity (99.3%) for consistent experimental results.
  • Soluble in DMSO at ≥50 mg/mL; several in vivo vehicle formulations reported.
  • Recommended storage supports long-term stability of powder and solutions.
  • Available in small research quantities for dosing flexibility.

Catalog No. 50-002-46742 Supplier Medchemexpress LLC Supplier No. HY1019810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Navarixin is an orally active, allosteric antagonist of CXCR1 and CXCR2 used as a research compound to study chemokine receptor signaling, neutrophil recruitment, and inflammatory responses in vitro and in vivo.

  • Potent, allosteric antagonist of CXCR1 and CXCR2.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High purity (99.3%) for consistent experimental results.
  • Soluble in DMSO at ≥50 mg/mL; several in vivo vehicle formulations reported.
  • Recommended storage supports long-term stability of powder and solutions.
  • Available in small research quantities for dosing flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.