Learn More
Cayman Chemical NavenI CD8MHC-I Ato647N 100RXN
SDP
Catalog No. 502945907
Encompass_Preferred
A KAT8 inhibitor selective for KAT8 over KAT2A KAT2B KAT3B KAT6A and KAT7 at 200 UM reduces H4K16Ac in HT-29 cells at 50 UM reduces the viability of HCT116 H1299 A549 and U937 cancer cells (IC50s 39.4 52.1 41 and 30.1 UM respectively)
Catalog No. 50-294-5907 Supplier Cayman Chemical Supplier No. 40924100 RXN
A KAT8 inhibitor selective for KAT8 over KAT2A KAT2B KAT3B KAT6A and KAT7 at 200 UM reduces H4K16Ac in HT-29 cells at 50 UM reduces the viability of HCT116 H1299 A549 and U937 cancer cells (IC50s 39.4 52.1 41 and 30.1 UM respectively)