Learn More
Cayman Chemical NavenI TrIflex Cel MR 100RXN
SDP
Catalog No. 502917169
Encompass_Preferred
An inhibitor of SARS-CoV and SARS-CoV-2 Mpro (IC50s 0.03 and 0.038 UM respectively) reduces viral infectivity in SARS-CoV-infected Vero cells (EC50 6.9 UM)
Catalog No. 50-291-7169 Supplier Cayman Chemical Supplier No. 39217100 RXN
An inhibitor of SARS-CoV and SARS-CoV-2 Mpro (IC50s 0.03 and 0.038 UM respectively) reduces viral infectivity in SARS-CoV-infected Vero cells (EC50 6.9 UM)