Learn More
SAPPHIRE NORTH AMERICA Naveni Triflex Cell MR
Catalog No. NC2540348
Encompass
Detect and quantify total protein A, total protein B, and the interaction AB between the two proteins. The assay is designed to be used with a mouse and rabbit primary antibody pair
Catalog No. NC2540348 Supplier SAPPHIRE NORTH AMERICA Supplier No. TF.100.MR
Detect and quantify total protein A, total protein B, and the interaction AB between the two proteins. The assay is designed to be used with a mouse and rabbit primary antibody pair