Learn More
Cayman Chemical NaveniBright BOND RX HRP, 1 ea
SDP
Catalog No. 502943656
Encompass_Preferred
The NaveniBirght™ BOND RX HRP is an in situ proximity ligation assay kit using BOND RX/RXm Fully Automated Research Stainer. The kit has been validated on FFPE human tissue samples. The kit is based on the Naveni in situ proximity ligation technology with an automated workflow. It uses two Navenibodies linked to proprietary oligo arms. The two Navenibodies included in the kit will bind primary antibodies of mouse and rabbit origin. When Navenibodies are in close proximity, a strong and distinct signal is generated. Read more about Naveni technology. Included in the kit: • Anti-mouse Navenibody • Anti-rabbit Navenibody • Buffers for blocking and dilutions, and all detection reagents • Reagents sufficient for 30 FFPE tissue slides, including dead volumes Not included in the kit: • Leica Biosystems for BOND RX/RXm with standard accessories. • Mounting medium
Catalog No. 50-294-3656 Supplier Cayman Chemical Supplier No. 416501 EA
The NaveniBirght™ BOND RX HRP is an in situ proximity ligation assay kit using BOND RX/RXm Fully Automated Research Stainer. The kit has been validated on FFPE human tissue samples. The kit is based on the Naveni in situ proximity ligation technology with an automated workflow. It uses two Navenibodies linked to proprietary oligo arms. The two Navenibodies included in the kit will bind primary antibodies of mouse and rabbit origin. When Navenibodies are in close proximity, a strong and distinct signal is generated. Read more about Naveni technology. Included in the kit: • Anti-mouse Navenibody • Anti-rabbit Navenibody • Buffers for blocking and dilutions, and all detection reagents • Reagents sufficient for 30 FFPE tissue slides, including dead volumes Not included in the kit: • Leica Biosystems for BOND RX/RXm with standard accessories. • Mounting medium