Learn More
SAPPHIRE NORTH AMERICA NaveniBright HRP
Catalog No. NC2386489
Encompass
The NaveniBright HRP kit is used for chromogenic visualization of protein-protein interactions, post-translational modifications like phosphorylation, or to increase the specificity when localizing single proteins. This assay is designed to be used with primary antibody pairs, generated in mouse and rabbit respectively.
Catalog No. NC2386489 Supplier SAPPHIRE NORTH AMERICA Supplier No. NB.MR.HRP.100
The NaveniBright HRP kit is used for chromogenic visualization of protein-protein interactions, post-translational modifications like phosphorylation, or to increase the specificity when localizing single proteins. This assay is designed to be used with primary antibody pairs, generated in mouse and rabbit respectively.