Learn More
Cayman Chemical NaveniFlex Cell MR Atto647N, 100 RXN
SDP
Catalog No. 502918935
Encompass_Preferred
The NaveniFlex Cell MR Atto647N kit is an in situ proximity ligation assay kit optimized for cell samples.; The kit enables fluorescence visualization of protein-protein interactions or post-translational modifications such as phosphorylation and ensures high specificity.; 39219).; Compatible with traditional immunostaining equipment, the kit uses secondary antibody detection to produce an amplified signal in the form of a fluorescent dot, thus facilitating immediate quantification of the readout.; The kit is based on our Naveni™ Proximity Ligation Technology, with two Navenibodies conjugated to proprietary oligo arms.; The technology ensures specific and sensitive detection.; Included in the kit: • Anti-mouse Navenibody • Anti-rabbit Navenibody • Detection fluorophore; Atto647N • Buffers for blocking and dilutions, and all detection reagents • The Navenibody solution has a working volume of 4000 µl.
Catalog No. 50-291-8935 Supplier Cayman Chemical Supplier No. 39504100 RXN
The NaveniFlex Cell MR Atto647N kit is an in situ proximity ligation assay kit optimized for cell samples.; The kit enables fluorescence visualization of protein-protein interactions or post-translational modifications such as phosphorylation and ensures high specificity.; 39219).; Compatible with traditional immunostaining equipment, the kit uses secondary antibody detection to produce an amplified signal in the form of a fluorescent dot, thus facilitating immediate quantification of the readout.; The kit is based on our Naveni™ Proximity Ligation Technology, with two Navenibodies conjugated to proprietary oligo arms.; The technology ensures specific and sensitive detection.; Included in the kit: • Anti-mouse Navenibody • Anti-rabbit Navenibody • Detection fluorophore; Atto647N • Buffers for blocking and dilutions, and all detection reagents • The Navenibody solution has a working volume of 4000 µl.