Learn More
Cayman Chemical NavenIFlex Cel MR RED 100RXN
SDP
Catalog No. 502919194
Encompass_Preferred
A DUSP26 inhibitor (IC50 11.62 UM) induces IMR-32 neuroblastoma cell apoptosis (IC50 4.13 UM)
Catalog No. 50-291-9194 Supplier Cayman Chemical Supplier No. 39505100 RXN
A DUSP26 inhibitor (IC50 11.62 UM) induces IMR-32 neuroblastoma cell apoptosis (IC50 4.13 UM)