Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Cell GR Atto647N
Catalog No. NC2540265
Encompass
Detect proteins, protein-protein interactions, or protein modifications in cell samples. The assay is designed to be used with a goat and rabbit primary antibody pair. Detection with Atto647N fluorophore is included.
Catalog No. NC2540265 Supplier SAPPHIRE NORTH AMERICA Supplier No. NC.GR.100.ATTO
Detect proteins, protein-protein interactions, or protein modifications in cell samples. The assay is designed to be used with a goat and rabbit primary antibody pair. Detection with Atto647N fluorophore is included.