Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Cell MR RED
Catalog No. NC2540262
Encompass
Detect proteins, protein-protein interactions, or protein modifications in cell samples. The assay is designed to be used with a mouse and rabbit primary antibody pair. Detection with TEX615 fluorophore is included.
Catalog No. NC2540262 Supplier SAPPHIRE NORTH AMERICA Supplier No. NC.MR.100.RED
Detect proteins, protein-protein interactions, or protein modifications in cell samples. The assay is designed to be used with a mouse and rabbit primary antibody pair. Detection with TEX615 fluorophore is included.