Learn More
Cayman Chemical NavenIFlex Control KIt 6RXN
SDP
Catalog No. 502916545
Encompass_Preferred
The NaveniFlex control kit has been designed as a control for the NaveniFlex MR proximity ligation reaction and to help the user distinguish between positive and negative proximity ligation signals. The kit is based on our Naveni™ Proximity Ligation Technology, with two Navenibodies conjugated to proprietary oligo arms. The technology ensures specific and sensitive detection; learn more about Naveni technology. Included in the kit: • Anti-mouse HER2 primary antibody • Anti-rabbit HER2 primary antibody • Three slides with fixed, pre-treated, and dehydrated BT474 cells • Control kit is ready to be used with the NaveniFlex MR protocol To be used with the NaveniFlex MR kit
Catalog No. 50-291-6545 Supplier Cayman Chemical Supplier No. 392056 RXN
The NaveniFlex control kit has been designed as a control for the NaveniFlex MR proximity ligation reaction and to help the user distinguish between positive and negative proximity ligation signals. The kit is based on our Naveni™ Proximity Ligation Technology, with two Navenibodies conjugated to proprietary oligo arms. The technology ensures specific and sensitive detection; learn more about Naveni technology. Included in the kit: • Anti-mouse HER2 primary antibody • Anti-rabbit HER2 primary antibody • Three slides with fixed, pre-treated, and dehydrated BT474 cells • Control kit is ready to be used with the NaveniFlex MR protocol To be used with the NaveniFlex MR kit