Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Tissue MR Atto647N
Catalog No. NC2540269
Encompass
NaveniFlex Tissue MR Atto647N is designed specifically for the detection of proteins, protein interactions or modification in tissues. It improves sensitivity and removes high background seen in certain samples. NaveniFlex Tissue MR Atto647N is compatible with a mouse and rabbit primary antibody pair and uses a Atto647N fluorophore for detection.
Catalog No. NC2540269 Supplier SAPPHIRE NORTH AMERICA Supplier No. NT.MR.100.ATTO
NaveniFlex Tissue MR Atto647N is designed specifically for the detection of proteins, protein interactions or modification in tissues. It improves sensitivity and removes high background seen in certain samples. NaveniFlex Tissue MR Atto647N is compatible with a mouse and rabbit primary antibody pair and uses a Atto647N fluorophore for detection.