Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Tissue MR RED
Catalog No. NC2540268
Encompass
NaveniFlex Tissue MR RED is designed specifically for the detection of proteins, protein interactions or modification in tissues. It improves sensitivity and removes high background seen in certain samples. NaveniFlex Tissue MR Red is compatible with a mouse and rabbit primary antibody pair and uses a TEX615 fluorophore for detection.
Catalog No. NC2540268 Supplier SAPPHIRE NORTH AMERICA Supplier No. NT.MR.100.RED
NaveniFlex Tissue MR RED is designed specifically for the detection of proteins, protein interactions or modification in tissues. It improves sensitivity and removes high background seen in certain samples. NaveniFlex Tissue MR Red is compatible with a mouse and rabbit primary antibody pair and uses a TEX615 fluorophore for detection.