Learn More
Medchemexpress LLC HY-10087 10mg Medchemexpress, Navitoclax CAS:923564-51-6 Purity:>98%
SDP
Catalog No. 501931976
Encompass_Preferred
Medchemexpress, HY-10087 10mg Navitoclax CAS:923564-51-6 Navitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-x L , Bcl-2 and Bcl-w, with a K i of less than 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1976 Supplier Medchemexpress LLC Supplier No. HY1008710MG
Medchemexpress, HY-10087 10mg Navitoclax CAS:923564-51-6 Navitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-x L , Bcl-2 and Bcl-w, with a K i of less than 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.