Learn More
TARGETMOL CHEMICALS INC Navitoclax
SDP
Supplier:  TARGETMOL CHEMICALS INC T2101200MG
Encompass_Preferred
Navitoclax (ABT-263) is a potent, orally bioavailable Bcl-2 family protein inhibitor that avidly binds Bcl-2, Bcl-xL, and Bcl-W (Ki < 1 nM for all three proteins).
Catalog No. NC1731582