Learn More
Medchemexpress LLC Navitoclax (ABT-263) | 923564-51-6 | MFCD12756219 | ≥98.0% | 974.61 g·mol⁻¹ | C47H55ClF3N5O6S3 | 200 MG
SDP
Catalog No. 5000211728
Encompass_Preferred
Navitoclax (ABT-263) is a potent, orally active small-molecule inhibitor of anti-apoptotic Bcl-2 family proteins used in preclinical apoptosis and oncology research. Supplied as a powder, it is used for both in vitro assays and in vivo models, with documented solubility and recommended storage for stability.
  • Potent Bcl-2 family inhibitor for apoptosis research.
  • Orally active small molecule suitable for in vivo studies.
  • High solubility in DMSO for convenient stock solutions.
  • Available in multiple quantities for dosing flexibility.
  • Recommended storage conditions to preserve stability.
Catalog No. 50-002-11728 Supplier Medchemexpress LLC Supplier No. HY10087200MG
Navitoclax (ABT-263) is a potent, orally active small-molecule inhibitor of anti-apoptotic Bcl-2 family proteins used in preclinical apoptosis and oncology research. Supplied as a powder, it is used for both in vitro assays and in vivo models, with documented solubility and recommended storage for stability.
  • Potent Bcl-2 family inhibitor for apoptosis research.
  • Orally active small molecule suitable for in vivo studies.
  • High solubility in DMSO for convenient stock solutions.
  • Available in multiple quantities for dosing flexibility.
  • Recommended storage conditions to preserve stability.