Learn More
Medchemexpress LLC Navitoclax-d8 | 1217620-38-6 | 99.7% | 982.66 | C47H47D8ClF3N5O6S3 | 1 MG
SDP
Catalog No. 5000211788
Encompass_Preferred
Navitoclax-d8 is the deuterium-labeled analog of navitoclax (ABT-263), a potent inhibitor of anti-apoptotic Bcl-2 family proteins. Supplied as a 1 mg stable-isotope reference, it is intended for use in mass spectrometry, pharmacokinetic, and mechanistic research where isotopic tracing or internal standards are required.
  • Deuterium-labeled (d8) analog for clear mass shift in MS analyses.
  • High purity offering reliable analytical performance.
  • Suitable as an internal standard for pharmacokinetic studies.
  • Supplied in a low-mass format for specialized research applications.
  • Stable under recommended storage for long-term use.
Catalog No. 50-002-11788 Supplier Medchemexpress LLC Supplier No. HY10087S1MG
Navitoclax-d8 is the deuterium-labeled analog of navitoclax (ABT-263), a potent inhibitor of anti-apoptotic Bcl-2 family proteins. Supplied as a 1 mg stable-isotope reference, it is intended for use in mass spectrometry, pharmacokinetic, and mechanistic research where isotopic tracing or internal standards are required.
  • Deuterium-labeled (d8) analog for clear mass shift in MS analyses.
  • High purity offering reliable analytical performance.
  • Suitable as an internal standard for pharmacokinetic studies.
  • Supplied in a low-mass format for specialized research applications.
  • Stable under recommended storage for long-term use.