Learn More
Medchemexpress LLC Navitoclax-piperazine | 2143096-93-7 | 99.7% | 973.63 | 1 ML
SDP
Catalog No. 5000404771
Encompass_Preferred
Navitoclax-piperazine (ABT-263-piperazine) is an analog of Navitoclax and acts as a BCL-XL inhibitor. It serves as the ligand for the target protein of PROTAC DT2216. This compound, when combined with an E3 ubiquitin ligase VHL ligand, can be used to synthesize PROTAC DT2216. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Analog of Navitoclax
  • Acts as a BCL-XL inhibitor
  • Serves as a ligand for target proteins
  • Can be used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-004-04771 Supplier Medchemexpress LLC Supplier No. HY4443210MM/1ML
Navitoclax-piperazine (ABT-263-piperazine) is an analog of Navitoclax and acts as a BCL-XL inhibitor. It serves as the ligand for the target protein of PROTAC DT2216. This compound, when combined with an E3 ubiquitin ligase VHL ligand, can be used to synthesize PROTAC DT2216. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Analog of Navitoclax
  • Acts as a BCL-XL inhibitor
  • Serves as a ligand for target proteins
  • Can be used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system for selective protein degradation