Learn More
Medchemexpress LLC Navlimetostat hydrochloride | 00-00-0 | 99.2% | C23H19Cl2FN6O2 | 10MG
SDP
Catalog No. 5000259650
Encompass_Preferred
Navlimetostat hydrochloride is a research-grade small-molecule inhibitor selective for the PRMT5-MTA complex. It demonstrates potent biochemical and cellular activity and is used in preclinical studies to probe PRMT5 biology and evaluate antineoplastic effects, with reported selectivity in MTAP-deleted models.
  • Potent inhibitor of the PRMT5-MTA complex (IC50 3.6 nM).
  • Inhibits PRMT5 enzymatic activity (IC50 20.5 nM).
  • Shows cellular selectivity in MTAP-deleted models (cell viability 12 nM vs 890 nM in parental cells).
  • High purity (99.2%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Suitable for preclinical research; store sealed away from moisture.
Catalog No. 50-002-59650 Supplier Medchemexpress LLC Supplier No. HY139611C10MG
Navlimetostat hydrochloride is a research-grade small-molecule inhibitor selective for the PRMT5-MTA complex. It demonstrates potent biochemical and cellular activity and is used in preclinical studies to probe PRMT5 biology and evaluate antineoplastic effects, with reported selectivity in MTAP-deleted models.
  • Potent inhibitor of the PRMT5-MTA complex (IC50 3.6 nM).
  • Inhibits PRMT5 enzymatic activity (IC50 20.5 nM).
  • Shows cellular selectivity in MTAP-deleted models (cell viability 12 nM vs 890 nM in parental cells).
  • High purity (99.2%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Suitable for preclinical research; store sealed away from moisture.