Learn More
Medchemexpress LLC Navlimetostat hydrochloride | 99.6% | 501.34 | 25 MG
SDP
Catalog No. 5000349926
Encompass_Preferred
Navlimetostat hydrochloride is a potent, orally active, and selective PRMT5-MTA complex inhibitor. It exhibits antineoplastic activity both in vitro and in vivo, making it a valuable compound for cancer studies.
  • Potent, orally active, selective inhibitor of the PRMT5-MTA complex
  • IC50s are 3.6 nM for PRMT5-MTA and 20.5 nM for PRMT5
  • Binds to the PRMT5-MTA complex with a KD value of 0.14 pM
  • Demonstrates antineoplastic activity in both in vitro and in vivo models
  • Used in research for cancer studies
  • Inhibits PRMT5 activity in MTAP-deleted HCT116 cells
  • Inhibits cell viability in MTAP del HCT116 cells
Catalog No. 50-003-49926 Supplier Medchemexpress LLC Supplier No. 25MGHY139611C
Navlimetostat hydrochloride is a potent, orally active, and selective PRMT5-MTA complex inhibitor. It exhibits antineoplastic activity both in vitro and in vivo, making it a valuable compound for cancer studies.
  • Potent, orally active, selective inhibitor of the PRMT5-MTA complex
  • IC50s are 3.6 nM for PRMT5-MTA and 20.5 nM for PRMT5
  • Binds to the PRMT5-MTA complex with a KD value of 0.14 pM
  • Demonstrates antineoplastic activity in both in vitro and in vivo models
  • Used in research for cancer studies
  • Inhibits PRMT5 activity in MTAP-deleted HCT116 cells
  • Inhibits cell viability in MTAP del HCT116 cells