Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Navocaftor | 2159103-66-7 | 99.2% | 416.33 | 10 MG
SDP

Catalog No. 5000344915
Encompass_Preferred

Navocaftor is a protein modulator that acts as a cystic fibrosis transmembrane regulator (CFTR). It is intended for research use only. This compound has a high purity and exhibits specific solubility characteristics for various research applications.

  • Solid appearance
  • Light yellow to yellow color
  • Purity of 99.20%
  • Stable for 3 years at -20°C as powder
  • Soluble in 10% DMSO, 90% corn oil for in vivo studies
  • Acts as a cystic fibrosis transmembrane regulator (CFTR) protein modulator

Catalog No. 50-003-44915 Supplier Medchemexpress LLC Supplier No. HY10915250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Navocaftor is a protein modulator that acts as a cystic fibrosis transmembrane regulator (CFTR). It is intended for research use only. This compound has a high purity and exhibits specific solubility characteristics for various research applications.

  • Solid appearance
  • Light yellow to yellow color
  • Purity of 99.20%
  • Stable for 3 years at -20°C as powder
  • Soluble in 10% DMSO, 90% corn oil for in vivo studies
  • Acts as a cystic fibrosis transmembrane regulator (CFTR) protein modulator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.