Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Navocaftor | 2159103-66-7 | 99.2% | 416.33 | 100 MG
SDP

Catalog No. 5000345472
Encompass_Preferred

Navocaftor (GLPG 3067) is a cystic fibrosis transmembrane regulator (CFTR) and a protein modulator.

  • Regulates cystic fibrosis transmembrane conductance.
  • Modulates protein function.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-45472 Supplier Medchemexpress LLC Supplier No. HY10915210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Navocaftor (GLPG 3067) is a cystic fibrosis transmembrane regulator (CFTR) and a protein modulator.

  • Regulates cystic fibrosis transmembrane conductance.
  • Modulates protein function.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.