Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Navocaftor | 2159103-66-7 | 99.2% | 416.33 | 50 MG
SDP

Catalog No. 5000343797
Encompass_Preferred

Navocaftor (GLPG 3067) is a protein modulator acting as a cystic fibrosis transmembrane regulator (CFTR), intended for research use only. Identified as a laboratory chemical for substance manufacturing, it has been tested and complies with specifications. Its chemical, physical, and toxicological properties are not fully investigated, and it is not validated for medical applications.

  • Acts as a cystic fibrosis transmembrane regulator (CFTR)
  • Functions as a protein modulator
  • For research use only
  • Solid, light yellow to yellow appearance
  • Purity of 99.2%
  • Molecular weight of 416.33

Catalog No. 50-003-43797 Supplier Medchemexpress LLC Supplier No. HY109152100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Navocaftor (GLPG 3067) is a protein modulator acting as a cystic fibrosis transmembrane regulator (CFTR), intended for research use only. Identified as a laboratory chemical for substance manufacturing, it has been tested and complies with specifications. Its chemical, physical, and toxicological properties are not fully investigated, and it is not validated for medical applications.

  • Acts as a cystic fibrosis transmembrane regulator (CFTR)
  • Functions as a protein modulator
  • For research use only
  • Solid, light yellow to yellow appearance
  • Purity of 99.2%
  • Molecular weight of 416.33

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.