Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Navocaftor | 2159103-66-7 | 99.2% | 416.33 g/mol | C15H11F3N4O5S | 5 MG
SDP

Catalog No. 5000264376
Encompass_Preferred

Navocaftor is a small-molecule cystic fibrosis transmembrane conductance regulator (CFTR) potentiator developed for research into restoring or enhancing CFTR channel function. It is supplied as a high-purity research reagent for in vitro and preclinical studies investigating CFTR modulation and combination therapy effects.

  • High purity (99.2%).
  • Molecular weight 416.3 g/mol.
  • Molecular formula C15H11F3N4O5S.
  • Intended for research use in CFTR modulation studies.
  • Supplied in 5 mg research quantities.

Catalog No. 50-002-64376 Supplier Medchemexpress LLC Supplier No. HY1091525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Navocaftor is a small-molecule cystic fibrosis transmembrane conductance regulator (CFTR) potentiator developed for research into restoring or enhancing CFTR channel function. It is supplied as a high-purity research reagent for in vitro and preclinical studies investigating CFTR modulation and combination therapy effects.

  • High purity (99.2%).
  • Molecular weight 416.3 g/mol.
  • Molecular formula C15H11F3N4O5S.
  • Intended for research use in CFTR modulation studies.
  • Supplied in 5 mg research quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.