Learn More
TARGETMOL CHEMICALS INC NAVTEMADLIN 5MG
SDP
Catalog No. 502578825
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Navtemadlin (AMG232) is a potent selective and orally available inhibitor of p53-MDM2 interaction (IC50 0.6 nM). It binds to MDM2 with a Kd of 0.045 nM. purity: 98%
Catalog No. 50-257-8825 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ01275MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Navtemadlin (AMG232) is a potent selective and orally available inhibitor of p53-MDM2 interaction (IC50 0.6 nM). It binds to MDM2 with a Kd of 0.045 nM. purity: 98%