Learn More
Braintree Scientific Navy Lysis Kit,1.5mL Eppendorf Safe-Lock tubes prefilled s/s
SDP
Supplier:  Braintree Scientific NAVYE1
Encompass
Navy Bead Lysis KitsExtra Power for your Tough SamplesNavy Bead Lysis Kits consist of microcentrifuge tubes pre-filled with a custom mix of stainless steel beads. They deliver extra homogenizing power for tough samples like bone. These kits provide: convenience; high yields with the multiple sizes of beads; and volumetric consistency of beads difficult to duplicate with manual dispensing of beads into tubes. Available in packs of 50 or 250, RNase-free or non-sterile. Eppendorf Navy kits are made with sturdy Eppendorf Safe-Lock snap-cap tubes. These 1.5 mL tubes are compatible with all microcentrifuge model Bullet Blenders.RINO Navy kits are made with our RINO screw-cap tubes, which are strong, durable, and contain an integrated rubber seal in the cap to provide an extra layer of protection against sample loss. These kits are for use with Storm and Gold microcentrifuge-model Bullet Blenders. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5342