Learn More
Next Advance Inc NAVY LYSIS KIT 250. BEAD LYSIS
SDP
Catalog No. NC1318781
Encompass
Navy Lysis Kit 250. Bead lysis kits for the homogenization of large, tough samples. Pack of 250, 1.5mL Eppendorf Safe-Lock tubes prefilled with stainless steel beads.
Catalog No. NC1318781 Supplier Next Advance Inc Supplier No. NAVYE5
Navy Lysis Kit 250. Bead lysis kits for the homogenization of large, tough samples. Pack of 250, 1.5mL Eppendorf Safe-Lock tubes prefilled with stainless steel beads.