Learn More
ABclonal Technology NAXE Rabbit pAb
SDP
Supplier:  ABclonal Technology A159521000UL
Encompass_Preferred
The product of this gene interacts with apolipoprotein A-I (apoA-I), the major apolipoprotein of high-density lipoproteins (HDLs). It is secreted into some bodily fluids, and its synthesis and secretion are stimulated in vitro by incubating cells with apoA-I. The human genome contains related pseudogenes.
Catalog No. 50-001-05585