Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NAZ2329 | 2809469-05-2 | 99.0% | 501.56 | C21H18F3NO4S3 | 10 MG
SDP

Catalog No. 5000245630
Encompass_Preferred

NAZ2329 is a cell-permeable small-molecule inhibitor of receptor-type protein tyrosine phosphatases PTPRZ and PTPRG supplied for research applications. It is characterized by micromolar inhibitory activity and is provided as a solid with recommended formulations for in vivo use.

  • CAS number 2809469-05-2; molecular formula C21H18F3NO4S3; molecular weight 501.56.
  • Purity 99.02% as supplied.
  • Targets PTPRZ (IC50 = 7.5 μM) and PTPRG (IC50 = 4.8 μM); shows activity against other PTP family members.
  • Solubility: DMSO 100 mg/mL; in vivo formulation examples and protocols available.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months).
  • For research use only; not for clinical use or sale to patients.

Catalog No. 50-002-45630 Supplier Medchemexpress LLC Supplier No. HY10369310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NAZ2329 is a cell-permeable small-molecule inhibitor of receptor-type protein tyrosine phosphatases PTPRZ and PTPRG supplied for research applications. It is characterized by micromolar inhibitory activity and is provided as a solid with recommended formulations for in vivo use.

  • CAS number 2809469-05-2; molecular formula C21H18F3NO4S3; molecular weight 501.56.
  • Purity 99.02% as supplied.
  • Targets PTPRZ (IC50 = 7.5 μM) and PTPRG (IC50 = 4.8 μM); shows activity against other PTP family members.
  • Solubility: DMSO 100 mg/mL; in vivo formulation examples and protocols available.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months).
  • For research use only; not for clinical use or sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.