Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naz2329 | 2809469-05-2 | 99.0% | 501.56 g·mol⁻¹ | C21H18F3NO4S3 | 50 MG
SDP

Catalog No. 5000245681
Encompass_Preferred

NAZ2329 is a small-molecule research inhibitor that allosterically and preferentially inhibits receptor-type protein tyrosine phosphatases PTPRZ and PTPRG. It is supplied as a solid for laboratory research and is characterized by high purity and established solubility in DMSO.

  • Allosterically inhibits PTPRZ and PTPRG (reported IC50: PTPRZ 7.5 μM, PTPRG 4.8 μM).
  • Cas number 2809469-05-2.
  • Purity 99.02%.
  • Molecular formula C21H18F3NO4S3.
  • Molecular weight 501.56 g·mol⁻¹.
  • Appearance solid, white to off-white.
  • Soluble in DMSO (~100 mg/mL).
  • Pack size 50 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-45681 Supplier Medchemexpress LLC Supplier No. HY10369350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NAZ2329 is a small-molecule research inhibitor that allosterically and preferentially inhibits receptor-type protein tyrosine phosphatases PTPRZ and PTPRG. It is supplied as a solid for laboratory research and is characterized by high purity and established solubility in DMSO.

  • Allosterically inhibits PTPRZ and PTPRG (reported IC50: PTPRZ 7.5 μM, PTPRG 4.8 μM).
  • Cas number 2809469-05-2.
  • Purity 99.02%.
  • Molecular formula C21H18F3NO4S3.
  • Molecular weight 501.56 g·mol⁻¹.
  • Appearance solid, white to off-white.
  • Soluble in DMSO (~100 mg/mL).
  • Pack size 50 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.