Learn More
Medchemexpress LLC NB-598 | 131060-14-5 | 449.67 | 100 MG
SDP
Catalog No. 5000395127
Encompass_Preferred
HY-16343, also known as NB-598, is identified as a potent and competitive inhibitor of squalene epoxidase (SE). It functions by suppressing triglyceride biosynthesis via the farnesol pathway. Furthermore, NB-598 serves as a click chemistry reagent, featuring an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups.
  • Potent and competitive inhibitor of squalene epoxidase (SE).
  • Suppresses triglyceride biosynthesis through the farnesol pathway.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group.
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
Catalog No. 50-003-95127 Supplier Medchemexpress LLC Supplier No. HY16343100MG
HY-16343, also known as NB-598, is identified as a potent and competitive inhibitor of squalene epoxidase (SE). It functions by suppressing triglyceride biosynthesis via the farnesol pathway. Furthermore, NB-598 serves as a click chemistry reagent, featuring an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups.
  • Potent and competitive inhibitor of squalene epoxidase (SE).
  • Suppresses triglyceride biosynthesis through the farnesol pathway.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group.
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.