Learn More
TARGETMOL CHEMICALS INC NB-598 2MG
SDP
Catalog No. 502877296
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. NB-598 is an effective and competitive inhibitor of squalene epoxidase. It suppresses triglyceride biosynthesis through the farnesol pathway. Purity 99.4%
Catalog No. 50-287-7296 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ01152MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. NB-598 is an effective and competitive inhibitor of squalene epoxidase. It suppresses triglyceride biosynthesis through the farnesol pathway. Purity 99.4%