Learn More
Medchemexpress LLC NB-598 hydrochloride | 136719-25-0 | 486.13 | 50 MG
SDP
Catalog No. 5000394172
Encompass_Preferred
NB-598 hydrochloride is a potent and competitive inhibitor of squalene epoxidase (SE), which suppresses triglyceride biosynthesis through the farnesol pathway. It is also a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent and competitive inhibitor of squalene epoxidase (SE).
  • Suppresses triglyceride biosynthesis via the farnesol pathway.
  • Functions as a click chemistry reagent, featuring an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.
Catalog No. 50-003-94172 Supplier Medchemexpress LLC Supplier No. HY1634350MG
NB-598 hydrochloride is a potent and competitive inhibitor of squalene epoxidase (SE), which suppresses triglyceride biosynthesis through the farnesol pathway. It is also a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent and competitive inhibitor of squalene epoxidase (SE).
  • Suppresses triglyceride biosynthesis via the farnesol pathway.
  • Functions as a click chemistry reagent, featuring an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.