Learn More
Medchemexpress LLC NB-598 maleate | 131060-14-5 | MFCD00898838 | 99.4% | 449.7 g/mol | C27H31NOS2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1634310MG
Encompass_Preferred
NB-598 is a research chemical inhibitor of squalene epoxidase used to modulate sterol biosynthesis in biochemical and pharmacological studies. It is supplied in small quantities for laboratory research and is commonly used to study cholesterol and triglyceride metabolism.
  • Potent competitive inhibitor of squalene epoxidase
  • Useful for studying sterol and triglyceride biosynthesis pathways
  • Available in small research-scale quantities suitable for in vitro and in vivo studies
  • High purity (reported ~99.4%)
  • Molecular formula C27H31NOS2; molecular weight 449.7 g/mol
Catalog No. 50-002-89861