Learn More
Medchemexpress LLC NB-598 Maleate | 155294-62-5 | 99.4% | 565.74 | 2 MG
SDP
Catalog No. 5000394729
Encompass_Preferred
NB-598 Maleate is a potent and competitive inhibitor of squalene epoxidase (SE) that suppresses triglyceride biosynthesis through the farnesol pathway. It also functions as a click chemistry reagent, containing an Alkyne group.
  • Potent and competitive inhibitor of squalene epoxidase (SE)
  • Suppresses triglyceride biosynthesis through the farnesol pathway
  • Click chemistry reagent containing an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-94729 Supplier Medchemexpress LLC Supplier No. HY16343C2MG
NB-598 Maleate is a potent and competitive inhibitor of squalene epoxidase (SE) that suppresses triglyceride biosynthesis through the farnesol pathway. It also functions as a click chemistry reagent, containing an Alkyne group.
  • Potent and competitive inhibitor of squalene epoxidase (SE)
  • Suppresses triglyceride biosynthesis through the farnesol pathway
  • Click chemistry reagent containing an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups