Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NB-598 Maleate | 155294-62-5 | 99.4% | 565.74 | 25 MG
SDP

Catalog No. 5000395395
Encompass_Preferred

NB-598 Maleate | 155294-62-5 | 99.4% | 565.74 | 25 MG

Catalog No. 50-003-95395 Supplier Medchemexpress LLC Supplier No. 25MGHY16343C
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NB-598 Maleate | 155294-62-5 | 99.4% | 565.74 | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.