Learn More
Medchemexpress LLC NB-598 (Maleate) | 155294-62-5 | 99.4% | 565.74 | 50 MG
SDP
Catalog No. 5000394733
Encompass_Preferred
NB-598 Maleate is a potent and competitive inhibitor of squalene epoxidase (SE), and it suppresses triglyceride biosynthesis through the farnesol pathway. It is also a click chemistry reagent, containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Potent and competitive inhibitor of squalene epoxidase (SE)
  • Suppresses triglyceride biosynthesis through the farnesol pathway
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-94733 Supplier Medchemexpress LLC Supplier No. HY16343C50MG
NB-598 Maleate is a potent and competitive inhibitor of squalene epoxidase (SE), and it suppresses triglyceride biosynthesis through the farnesol pathway. It is also a click chemistry reagent, containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Potent and competitive inhibitor of squalene epoxidase (SE)
  • Suppresses triglyceride biosynthesis through the farnesol pathway
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups