Learn More
Medchemexpress LLC NB-598 (maleate) | 155294-62-5 | MFCD22665748 | 99.4% | 565.74 | C31H35NO5S2 | 10 MG
SDP
Catalog No. 5000289090
Encompass_Preferred
NB-598 maleate is a research-grade small molecule that acts as a potent, competitive inhibitor of squalene epoxidase (SE). It is used in biochemical and cell-based studies to suppress triglyceride and sterol biosynthesis via the farnesol pathway and is supplied as the maleate salt in solid form or as a concentrated DMSO solution for assay use.
  • Potent competitive inhibitor of squalene epoxidase.
  • Used to suppress triglyceride and cholesterol biosynthesis in biochemical studies.
  • Supplied as the maleate salt for improved stability and handling.
  • Available as small solid quantities and as a 10 mM solution in DMSO for convenience in assays.
  • High reported purity (~99.4%) suitable for research applications.
Catalog No. 50-002-89090 Supplier Medchemexpress LLC Supplier No. HY16343C10MG
NB-598 maleate is a research-grade small molecule that acts as a potent, competitive inhibitor of squalene epoxidase (SE). It is used in biochemical and cell-based studies to suppress triglyceride and sterol biosynthesis via the farnesol pathway and is supplied as the maleate salt in solid form or as a concentrated DMSO solution for assay use.
  • Potent competitive inhibitor of squalene epoxidase.
  • Used to suppress triglyceride and cholesterol biosynthesis in biochemical studies.
  • Supplied as the maleate salt for improved stability and handling.
  • Available as small solid quantities and as a 10 mM solution in DMSO for convenience in assays.
  • High reported purity (~99.4%) suitable for research applications.