Learn More
New England Biolabs, Inc. Nb.BbvCI – 1000 units
SDP
Catalog No. 50812376
Encompass
A restriction endonuclease that recognizes the sequence CCTCA_GC.
Catalog No. 50-812-376 Supplier New England Biolabs, Inc. Supplier No. R0631S
A restriction endonuclease that recognizes the sequence CCTCA_GC.