Learn More
New England Biolabs, Inc. Nb.BsmI – 1000 units
SDP
Catalog No. 50995280
Encompass
A restriction endonuclease that recognizes the sequence GAATG_CN.
Catalog No. 50-995-280 Supplier New England Biolabs, Inc. Supplier No. R0706S
A restriction endonuclease that recognizes the sequence GAATG_CN.