Learn More
New England Biolabs, Inc. Nb.BsrDI – 5000 units
SDP
Catalog No. 50995271
Encompass
A restriction endonuclease that recognizes the sequence GCAATG_NN.
Catalog No. 50-995-271 Supplier New England Biolabs, Inc. Supplier No. R0648L
A restriction endonuclease that recognizes the sequence GCAATG_NN.